Learn More
Medchemexpress LLC 4-Thiouridine 5'-triphosphate tetralithium | 20188-72-1 | C9H11Li4N2O14P3S | 25 MG
SDP
Catalog No. 5000374937
Encompass_Preferred
4-Thiouridine 5'-triphosphate tetralithium (4-Thio-UTP tetralithium) is a UTP analog and a potent P2Y2 and P2Y4 agonist, with EC50 values of 35 nM for hP2Y2 and 350 nM for hP2Y4. This laboratory chemical is suitable for cross-linking experiments and transcriptional complex labeling studies. It is for research use only and has not been fully validated for medical applications.
  • Potent P2Y2 and P2Y4 agonist
  • EC50 values of 35 nM for hP2Y2 and 350 nM for hP2Y4
  • Suitable for cross-linking experiments
  • Useful in transcriptional complex labeling studies
Catalog No. 50-003-74937 Supplier Medchemexpress LLC Supplier No. 25MGHY154840
4-Thiouridine 5'-triphosphate tetralithium (4-Thio-UTP tetralithium) is a UTP analog and a potent P2Y2 and P2Y4 agonist, with EC50 values of 35 nM for hP2Y2 and 350 nM for hP2Y4. This laboratory chemical is suitable for cross-linking experiments and transcriptional complex labeling studies. It is for research use only and has not been fully validated for medical applications.
  • Potent P2Y2 and P2Y4 agonist
  • EC50 values of 35 nM for hP2Y2 and 350 nM for hP2Y4
  • Suitable for cross-linking experiments
  • Useful in transcriptional complex labeling studies