Learn More
Selleck Chemical LLC 4-Thiouridine-E1292-1G
SDP
Catalog No. 503134988
Encompass_Preferred
4-thiouridine (4-Thio-UTP) a thiouridine and a nucleoside analogue has a role as an affinity label and an antimetabolite
Catalog No. 50-313-4988 Supplier Selleck Chemical LLC Supplier No. E12921G
4-thiouridine (4-Thio-UTP) a thiouridine and a nucleoside analogue has a role as an affinity label and an antimetabolite