Learn More
Abcam 4-Thiouridine, RNA synthesis inhibitor, 5MG
Catalog No. 502583292
Encompass_Preferred
MW 260.27 Da, Purity >99%. Ribonucleoside (Uridine ab143255) analog. Antisense agent. RNA synthesis inhibitor. Modifies oligos slated for RNA. Induces p53 and inhibits proliferation. Metabolically labels RNA for stability studies.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-3292 Supplier Abcam Supplier No. AB1437185MG
MW 260.27 Da, Purity >99%. Ribonucleoside (Uridine ab143255) analog. Antisense agent. RNA synthesis inhibitor. Modifies oligos slated for RNA. Induces p53 and inhibits proliferation. Metabolically labels RNA for stability studies.
The product is subject to the following: Abcam Restricted Use Statement