Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(trans-4-Butylcyclohexyl)cyclohexanone 98.0+%, TCI America™
SDP

Catalog No. B49465G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49465G 5 g
B494625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B49465G Supplier TCI America Supplier No. B49465G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 92413-47-3
Molecular Formula C16H28O
Molecular Weight (g/mol) 236.399
MDL Number MFCD06797514
InChI Key LDUYZTOYURUHIX-UHFFFAOYSA-N
PubChem CID 23440378
IUPAC Name 4-(4-butylcyclohexyl)cyclohexan-1-one
SMILES CCCCC1CCC(CC1)C2CCC(=O)CC2

Specifications

Melting Point 44°C
Color White
Quantity 5 g
Formula Weight 236.40
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(trans-4-Butylcyclohexyl)cyclohexanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.