Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(trans-4-Pentylcyclohexyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. P23631G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2363-1G 1 g
P23635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P2363-1G Supplier TCI America Supplier No. P23631G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 143651-26-7
Molecular Formula C17H27BO2
MDL Number MFCD09750938
InChI Key JFESOTHKCUMHGA-UHFFFAOYSA-N
Synonym 4-(trans-4-Pentylcyclohexyl)benzeneboronic Acid
PubChem CID 3696091
IUPAC Name [4-(4-pentylcyclohexyl)phenyl]boronic acid

Specifications

Color White
Quantity 1 g
SMILES CCCCCC1CCC(CC1)C1=CC=C(C=C1)B(O)O
Molecular Weight (g/mol) 274.21
Formula Weight 274.21
Physical Form Crystalline Powder
Chemical Name or Material 4-(trans-4-Pentylcyclohexyl)phenylboronic Acid (contains varying amounts of Anhydride)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.