Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(trans-4-Propylcyclohexyl)benzonitrile 98.0+%, TCI America™
SDP

Catalog No. P16175G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P16175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P16175G Supplier TCI America Supplier No. P16175G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 61203-99-4
Molecular Formula C16H21N
Molecular Weight (g/mol) 227.351
MDL Number MFCD04038799
InChI Key XXUSEPRYHRDKFV-UHFFFAOYSA-N
Synonym 4-4-propylcyclohexyl benzonitrile, 4-trans-4-propylcyclohexyl benzonitrile, trans-4-4-propylcyclohexyl benzonitrile, 1-cyano-4-trans-4-propylcyclohexyl benzene, 4-trans-4'-n-propylcyclohexyl benzonitrile, benzonitrile, 4-trans-4-propylcyclohexyl, 4-4-propylcyclohexyl benzenecarbonitrile, 4-1s,4r-4-propylcyclohexyl benzonitrile, trans-4-propyl 4-cyanophenyl cyclohexane
PubChem CID 109061
IUPAC Name 4-(4-propylcyclohexyl)benzonitrile
SMILES CCCC1CCC(CC1)C2=CC=C(C=C2)C#N

Specifications

Melting Point 45°C
Color White-Yellow
Quantity 5 g
UN Number 3439
Formula Weight 227.35
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(trans-4-Propylcyclohexyl)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.