Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(trans-4-Propylcyclohexyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. P21625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2162-5G 5 g
P21621G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P2162-5G Supplier TCI America Supplier No. P21625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 146862-02-4
Molecular Formula C15H23BO2
MDL Number MFCD06658197
InChI Key QTBUVZSHCNAPRT-UHFFFAOYSA-N
PubChem CID 3104710
IUPAC Name [4-(4-propylcyclohexyl)phenyl]boronic acid

Specifications

Color White
Quantity 5 g
Synonym 4-trans-4-propylcyclohexyl phenylboronic acid, 4-trans-4-n-propylcyclohexyl phenyl boronic acid, 4-trans-4-propylcyclohexyl phenyl boronic acid, 4-4-propylcyclohexyl phenyl boronic acid, 4-4-propylcyclohexyl phenylboronic acid
SMILES CCCC1CCC(CC1)C1=CC=C(C=C1)B(O)O
Molecular Weight (g/mol) 246.16
Formula Weight 246.16
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.