Learn More
Medchemexpress LLC 4-trans-hydroxy-L-proline methyl ester hydrochloride | 40216-83-9 | MFCD00080855 | 97.0% | 181.62 | C6H12ClNO3 | 100 G
SDP
Catalog No. 5000202348
Encompass_Preferred
H-Hyp-OMe hydrochloride is the hydrochloride salt of a hydroxyproline methyl ester used as a small-molecule building block for linker synthesis in antibody-drug conjugate (ADC) and proteolysis targeting chimera (PROTAC) projects. It is supplied as a stable hydrochloride salt for bench-scale synthesis and linker incorporation.
  • Hydrochloride salt form for improved stability.
  • Non-cleavable linker building block for ADC synthesis.
  • Suitable as an alkyl chain-based PROTAC linker component.
  • Molecular formula: C6H12ClNO3; molecular weight: 181.62.
  • Available in bulk laboratory pack sizes, including 100 g.
Catalog No. 50-002-02348 Supplier Medchemexpress LLC Supplier No. HY76043100G
H-Hyp-OMe hydrochloride is the hydrochloride salt of a hydroxyproline methyl ester used as a small-molecule building block for linker synthesis in antibody-drug conjugate (ADC) and proteolysis targeting chimera (PROTAC) projects. It is supplied as a stable hydrochloride salt for bench-scale synthesis and linker incorporation.
  • Hydrochloride salt form for improved stability.
  • Non-cleavable linker building block for ADC synthesis.
  • Suitable as an alkyl chain-based PROTAC linker component.
  • Molecular formula: C6H12ClNO3; molecular weight: 181.62.
  • Available in bulk laboratory pack sizes, including 100 g.