Learn More
Medchemexpress LLC 4-trans-hydroxy-L-proline methyl ester hydrochloride | 40216-83-9 | MFCD00080855 | ≥98.0% | 181.62 | C6H12ClNO3 | 250 G
SDP
Catalog No. 5000202910
Encompass_Preferred
H-Hyp-OMe hydrochloride is the hydrochloride salt of 4-trans-hydroxy-L-proline methyl ester, supplied as a research reagent for use in organic synthesis and linker chemistry. It is typically used in antibody-drug conjugate (ADC) linker assembly and PROTAC linker construction.
  • used as a non-cleavable ADC linker building block
  • used in PROTAC and linker chemistry
  • supplied as the hydrochloride salt for improved stability
  • typical purity ≥98.0% by HPLC
  • available in 100 g, 250 g, 500 g, and 1 kg pack sizes
Catalog No. 50-002-02910 Supplier Medchemexpress LLC Supplier No. HY76043250G
H-Hyp-OMe hydrochloride is the hydrochloride salt of 4-trans-hydroxy-L-proline methyl ester, supplied as a research reagent for use in organic synthesis and linker chemistry. It is typically used in antibody-drug conjugate (ADC) linker assembly and PROTAC linker construction.
  • used as a non-cleavable ADC linker building block
  • used in PROTAC and linker chemistry
  • supplied as the hydrochloride salt for improved stability
  • typical purity ≥98.0% by HPLC
  • available in 100 g, 250 g, 500 g, and 1 kg pack sizes