Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Trifluoromethoxy)anisole 97.0+%, TCI America™
SDP

Catalog No. T17795G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T17795G Supplier TCI America Supplier No. T17795G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 710-18-9
Molecular Formula C8H7F3O2
Molecular Weight (g/mol) 192.137
MDL Number MFCD00216942
InChI Key NOAFZIOGGDPYKK-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy anisole, 1-methoxy-4-trifluoromethoxy benzene, p-trifluoromethoxy anisole, 4-trifluoromethoxyanisole, benzene, 1-methoxy-4-trifluoromethoxy, 4-methoxy trifluoromethoxy benzene, hydroquinone methyl trifluoromethyl ether, trifluoro 4-methoxyphenoxy methane, pubchem3570, acmc-1bcz3
PubChem CID 2724973
IUPAC Name 1-methoxy-4-(trifluoromethoxy)benzene
SMILES COC1=CC=C(C=C1)OC(F)(F)F

Specifications

Color Yellow
Quantity 5 g
UN Number 1993
Formula Weight 192.14
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-(Trifluoromethoxy)anisole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.