Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Trifluoromethoxy)benzenesulfonamide 98.0+%, TCI America™
SDP

Catalog No. T24275G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T24275G Supplier TCI America Supplier No. T24275G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 1513-45-7
Melting Point 149°C
Color White-Yellow
Molecular Formula C7H6F3NO3S
Quantity 5 g
MDL Number MFCD00798381
Synonym 4-trifluoromethoxy benzenesulfonamide, p-trifluoromethoxy benzenesulfonamide, 4-trifluoromethoxy benzenesulphonamide, 4-trifluoromethoxy benzene-1-sulfonamide, benzenesulfonamide, 4-trifluoromethoxy, pubchem11787, rgojchyybkmrll-uhfffaoysa, 4-trifluoromethoxybenzensulfonamide, trifluoromethoxy benzenesulfonamide, 4-trifluoromethoxybenzenesulphonyl amine
InChI Key RGOJCHYYBKMRLL-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1OC(F)(F)F)S(=O)(=O)N
IUPAC Name 4-(trifluoromethoxy)benzenesulfonamide
Molecular Weight (g/mol) 241.184
PubChem CID 2777211
Formula Weight 241.18
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Trifluoromethoxy)benzenesulfonamide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.