Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Trifluoromethoxy)benzoic acid, 97%, Thermo Scientific™

Catalog No. AAA1362114
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1362114 25 g
AAA1362103 1 g
AAA1362106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1362114 Supplier Thermo Scientific Chemicals Supplier No. A1362114

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 330-12-1
Molecular Formula C8H5F3O3
Molecular Weight (g/mol) 206.12
MDL Number MFCD00002541
InChI Key RATSANVPHHXDCT-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy benzoic acid, p-trifluoromethoxybenzoic acid, 4-trifluoromethoxy-benzoic acid, p-carboxyphenyl trifluoromethyl ether, 4-trifluoromethoxy benzoicacid, benzoic acid, 4-trifluoromethoxy, alpha,alpha,alpha-trifluoro-p-anisic acid, pubchem4593, acmc-1cp95, ksc225q9l
PubChem CID 67613
IUPAC Name 4-(trifluoromethoxy)benzoic acid
SMILES C1=CC(=CC=C1C(=O)O)OC(F)(F)F

Specifications

Melting Point 151°C to 154°C
Quantity 25 g
Beilstein 977356
Formula Weight 206.12
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethoxy)benzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 206-352-3
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.