Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-(trifluoromethoxy)-N-[[2-(trifluoromethyl)phenyl]methyl]-benzenecarboximidamide | 305339-41-7 | 99.8% | 1 ML
SDP

Catalog No. 5000232954
Encompass_Preferred

BZAD-01 is a potent, selective, orally active inhibitor of the NMDA receptor NR2B subunit (reported Ki = 72 nM). It is supplied as a solid (white to off-white) and as a 10 mM stock solution in DMSO.

  • Potent, selective NR2B inhibitor (Ki = 72 nM).
  • Molecular formula C16H12F6N2O; molecular weight 362.27.
  • High purity (approximately 99.8%).
  • Available as solid and as 10 mM solution in DMSO (1 mL).
  • Storage: solid at 4°C, protect from light; stock solutions -80°C up to 6 months or -20°C up to 1 month, protect from light.
  • Suitable for in vitro pharmacology and in vivo NR2B-related studies.

Catalog No. 50-002-32954 Supplier Medchemexpress LLC Supplier No. HY12110010MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BZAD-01 is a potent, selective, orally active inhibitor of the NMDA receptor NR2B subunit (reported Ki = 72 nM). It is supplied as a solid (white to off-white) and as a 10 mM stock solution in DMSO.

  • Potent, selective NR2B inhibitor (Ki = 72 nM).
  • Molecular formula C16H12F6N2O; molecular weight 362.27.
  • High purity (approximately 99.8%).
  • Available as solid and as 10 mM solution in DMSO (1 mL).
  • Storage: solid at 4°C, protect from light; stock solutions -80°C up to 6 months or -20°C up to 1 month, protect from light.
  • Suitable for in vitro pharmacology and in vivo NR2B-related studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.