Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-(Trifluoromethoxy)Phenylmagnesium Bromide Solution

Catalog No. 68790150ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
68-790-150ML 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 68-790-150ML Supplier Sigma Aldrich Supplier No. 68790150ML
Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant for:Preparation of chromanol derivatives as novel class of CETP inhibitors for treatment of cardiovascular diseasePd-catalyzed cross couplingPreparation of indazole-pyridine derivatives as protein kinase B/Akt inhibitors with reduced hypotension, via Stille coupling, Mitsunobu reaction and copper-catalyzed aziridine ring-opening reaction

Specifications

CAS 40438-64-0
Melting Point 103°C to 108°C
Molecular Formula C20H32NP
Linear Formula C20H32NP
MDL Number MFCD11044859
Quantity 1 g
Molecular Weight (g/mol) 317.45 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.