Learn More
Medchemexpress LLC 4-(Trifluoromethyl)-1H-pyrazole | 52222-73-8 | 136.08 | 1 ML
SDP
Catalog No. 5000445612
Encompass_Preferred
4-(Trifluoromethyl)-1H-pyrazole is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is also used as a pharmaceutical intermediate. It is for research use only and not sold to patients.
  • Used as pharmaceutical intermediates.
  • Palladium-catalyzed cross-coupling reactions of 5-tributylstannyl-4-fluoro-1H-pyrazoles with aryl iodides yielded high amounts of the corresponding 5-aryl-4-fluoro-1H-pyrazoles.
Catalog No. 50-004-45612 Supplier Medchemexpress LLC Supplier No. HYW00421210MM/1ML
4-(Trifluoromethyl)-1H-pyrazole is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is also used as a pharmaceutical intermediate. It is for research use only and not sold to patients.
  • Used as pharmaceutical intermediates.
  • Palladium-catalyzed cross-coupling reactions of 5-tributylstannyl-4-fluoro-1H-pyrazoles with aryl iodides yielded high amounts of the corresponding 5-aryl-4-fluoro-1H-pyrazoles.