Learn More
Medchemexpress LLC 4-(Trifluoromethyl)-1H-pyrazole | 52222-73-8 | 136.08 | 5 G
SDP
Catalog No. 5000447498
Encompass_Preferred
4-(Trifluoromethyl)-1H-pyrazole is a biochemical reagent and pharmaceutical intermediate. It is used as a biological material or organic compound for life science research. This product can be used in palladium-catalyzed cross-coupling reactions to provide high yields of corresponding 5-aryl-4-fluoro-1H-pyrazoles.
  • Biochemical reagent for life science research
  • Used as a biological material
  • Organic compound for various applications
  • Utilized in pharmaceutical intermediate synthesis
Catalog No. 50-004-47498 Supplier Medchemexpress LLC Supplier No. HYW0042125G
4-(Trifluoromethyl)-1H-pyrazole is a biochemical reagent and pharmaceutical intermediate. It is used as a biological material or organic compound for life science research. This product can be used in palladium-catalyzed cross-coupling reactions to provide high yields of corresponding 5-aryl-4-fluoro-1H-pyrazoles.
  • Biochemical reagent for life science research
  • Used as a biological material
  • Organic compound for various applications
  • Utilized in pharmaceutical intermediate synthesis