Learn More
Medchemexpress LLC 4'-Trifluoromethyl-2-biphenyl carboxylic acid | 84392-17-6 | 99.9% | 266.22 | 1 G
SDP
Catalog No. 5000404333
Encompass_Preferred
4'-Trifluoromethyl-2-biphenyl carboxylic acid is an orally active hypolipidemic agent. For research use only; not for sale to patients.
  • Orally active hypolipidemic agent.
  • Shows hypolipidemic activity in diet-induced hypercholesterolemic African green monkeys.
Catalog No. 50-004-04333 Supplier Medchemexpress LLC Supplier No. HY347811G
4'-Trifluoromethyl-2-biphenyl carboxylic acid is an orally active hypolipidemic agent. For research use only; not for sale to patients.
  • Orally active hypolipidemic agent.
  • Shows hypolipidemic activity in diet-induced hypercholesterolemic African green monkeys.