Learn More
Medchemexpress LLC 4'-Trifluoromethyl-2-biphenyl carboxylic acid | 84392-17-6 | 99.9% | 266.22 | 5 G
SDP
Catalog No. 5000404486
Encompass_Preferred
4'-Trifluoromethyl-2-biphenyl carboxylic acid is an orally active hypolipidemic agent, suitable for research purposes. It has demonstrated hypolipidemic activity in diet-induced hypercholesterolemic African green monkeys, leading to reduced serum LDL-cholesterol concentrations. This compound appears as a white to off-white solid.
  • Orally active hypolipidemic agent
  • Shows hypolipidemic activity in diet-induced hypercholesterolemic African green monkeys
  • Reduces serum LDL-cholesterol concentrations
  • White to off-white solid appearance
  • Soluble in DMSO (100 mg/mL)
  • Insoluble in H2O (< 0.1 mg/mL)
Catalog No. 50-004-04486 Supplier Medchemexpress LLC Supplier No. HY347815G
4'-Trifluoromethyl-2-biphenyl carboxylic acid is an orally active hypolipidemic agent, suitable for research purposes. It has demonstrated hypolipidemic activity in diet-induced hypercholesterolemic African green monkeys, leading to reduced serum LDL-cholesterol concentrations. This compound appears as a white to off-white solid.
  • Orally active hypolipidemic agent
  • Shows hypolipidemic activity in diet-induced hypercholesterolemic African green monkeys
  • Reduces serum LDL-cholesterol concentrations
  • White to off-white solid appearance
  • Soluble in DMSO (100 mg/mL)
  • Insoluble in H2O (< 0.1 mg/mL)