Learn More
Medchemexpress LLC 4-(Trifluoromethyl)aniline-d4 | 1219795-48-8 | 99.23% | C7H2D4F3N | 5 MG
SDP
Catalog No. 5000434962
Encompass_Preferred
4-(Trifluoromethyl)aniline-d4 is a deuterium-labeled derivative of 4-(Trifluoromethyl)aniline, which acts as a DYRK1A and DYRK1B kinase inhibitor. This compound incorporates stable heavy isotopes for use in various analytical applications.
  • Can be used as a tracer.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-34962 Supplier Medchemexpress LLC Supplier No. HY77324S5MG
4-(Trifluoromethyl)aniline-d4 is a deuterium-labeled derivative of 4-(Trifluoromethyl)aniline, which acts as a DYRK1A and DYRK1B kinase inhibitor. This compound incorporates stable heavy isotopes for use in various analytical applications.
  • Can be used as a tracer.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs.