Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Trifluoromethyl)aniline hydrochloride, 97%

Catalog No. AAA1832906
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1832906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1832906 Supplier Thermo Scientific Chemicals Supplier No. A1832906
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 90774-69-9 | C7H7ClF3N | 197.585 g/mol

4-(Trifluoromethyl)aniline hydrochloride is used in the synthesis of 2-(4-((6-Chloro-2-quinoxalinyl)oxy)phenoxy)-N-(4-(trifluoromethyl)phenyl)propanamide. It is also used to produce N-(p-trifluoromethylphenyl)phosphoramidic dichloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Trifluoromethyl)aniline hydrochloride is used in the synthesis of 2-(4-((6-Chloro-2-quinoxalinyl)oxy)phenoxy)-N-(4-(trifluoromethyl)phenyl)propanamide. It is also used to produce N-(p-trifluoromethylphenyl)phosphoramidic dichloride.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is hygroscopic in nature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90774-69-9
Molecular Formula C7H7ClF3N
Molecular Weight (g/mol) 197.585
MDL Number MFCD00007897
InChI Key CZGDYZDUMJDQSD-UHFFFAOYSA-N
Synonym 4-aminobenzotrifluoride hydrochloride, 4-trifluoromethyl aniline hydrochloride, 4-trifluoromethyl anilinium chloride, p-trifluoromethylaniline hydrochloride, 4-trifluoromethyl phenylamine, chloride, pubchem2816, 4-aminobenzotrifluoride, hydrochloride, 4-trifluoromethylaniline hydrochloride, 4-trifluoromethyl aniline;hydrochloride, a,a,a-trifluoro-p-toluidine hydrochloride
PubChem CID 2723961
IUPAC Name 4-(trifluoromethyl)aniline;hydrochloride
SMILES C1=CC(=CC=C1C(F)(F)F)N.Cl

Specifications

Melting Point ∼240°C (decomposition)
Quantity 5 g
UN Number UN2811
Beilstein 3629991
Formula Weight 197.59
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethyl)aniline hydrochloride
Hazard Category H302+H312+H332-H315-H319-H335-H373
Hazard Statement GHS H Statement
H301-H373-H315-H319-H335
Toxic if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 292-679-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.