Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Matrix Scientific 4-(Trifluoromethyl)benzaldehyde, 455-19-6, MFCD00006952, 100g

Catalog No. 500118120
Encompass_Preferred

Molecular Formula C8H5F3O, Purity 98%, Molecular Weight 174.12, Boiling Point 66-67/13mm

Catalog No. 50-011-8120 Supplier Matrix Scientific Supplier No. 005487100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Molecular Formula C8H5F3O, Purity 98%, Molecular Weight 174.12, Boiling Point 66-67/13mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.