Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-(Trifluoromethyl)benzaldehyde, 97%

Click to view available options
Quantity:
5g
25g
455-19-6
C8H5F3O
174.122
MFCD00006952
BEOBZEOPTQQELP-UHFFFAOYSA-N
4-trifluoromethyl benzaldehyde,benzaldehyde, 4-trifluoromethyl,p-trifluoromethylbenzaldehyde,p-trifluoromethyl benzaldehyde,alpha,alpha,alpha-trifluoro-p-tolualdehyde,4'-trifluoromethyl benzaldehyde,p-trifluoromethyl-benzaldhyde,4-trifluoromethyl-benzaldehyde
67996
4-(trifluoromethyl)benzaldehyde
C1=CC(=CC=C1C=O)C(F)(F)F
455-19-6
C8H5F3O
174.122
MFCD00006952
BEOBZEOPTQQELP-UHFFFAOYSA-N
4-trifluoromethyl benzaldehyde,benzaldehyde, 4-trifluoromethyl,p-trifluoromethylbenzaldehyde,p-trifluoromethyl benzaldehyde,alpha,alpha,alpha-trifluoro-p-tolualdehyde,4'-trifluoromethyl benzaldehyde,p-trifluoromethyl-benzaldhyde,4-trifluoromethyl-benzaldehyde
67996
4-(trifluoromethyl)benzaldehyde
C1=CC(=CC=C1C=O)C(F)(F)F
Applications
4-(Trifluoromethyl)benzaldehyde is useful reagent in kinetic studies of the asymmetric synthesis of alcohols and of the Wittig reaction. It is also used as Pharmaceutical intermediates.

Solubility
Soluble in water. 1.5 g/L at 20°C

Notes
Air Sensitive. Store away from air and oxidizing agents. Store under an inert atmosphere. Incompatible with oxidizing agents, reducing agents.
Density 1.275
Sensitivity Air Sensitive
Refractive Index 1.463
Flash Point 65°C (149°F)
Boiling Point 80°C to 81°C (25mmHg)
Melting Point 1°C to 2°C
Quantity 5g
Beilstein 1101680
Solubility Information Soluble in water. 1.5g/L at 20°C
Formula Weight 174.12
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethyl)benzaldehyde
Show More Show Less

GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H320-H335

EINECSNumber : 207-240-7

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon