Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Trifluoromethyl)benzene-1-sulfonyl chloride, 97%, Thermo Scientific™

Catalog No. TL00175EA
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
TL00175EA 10 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. TL00175EA Supplier Thermo Scientific Chemicals Supplier No. TL00175EA

Chemical Identifiers

CAS 2991-42-6
Molecular Formula C7H4ClF3O2S
Molecular Weight (g/mol) 244.61
MDL Number MFCD00042422
InChI Key OZDCZHDOIBUGAJ-UHFFFAOYSA-N
Synonym 4-trifluoromethyl benzenesulfonyl chloride, 4-trifluoromethyl benzene-1-sulfonyl chloride, 4-trifluoromethyl benzene sulfonyl chloride, p-trifluoromethylbenzenesulfonyl chloride, 4-trifluoromethyl benzenesulfonylchloride, 4-trifluoromethyl-benzenesulfonyl chloride, 4-trifluoromethyl phenylsulfonyl chloride, p-trifluoromethyl benzenesulfonyl chloride, 4-trifluoromethyl benzenesulphonyl chloride, benzenesulfonyl chloride, 4-trifluoromethyl
PubChem CID 2777399
IUPAC Name 4-(trifluoromethyl)benzenesulfonyl chloride
SMILES FC(F)(F)C1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula CF3OC6H4SO2Cl
Quantity 10 g
Formula Weight 244.62
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethyl)benzene-1-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.