Learn More
Medchemexpress LLC 4-(trifluoromethyl)benzoic acid-13C6 | 2731552-41-1 | 98.3% | 196.08 | C213C6H5F3O2 | 1 MG
SDP
Catalog No. 5000302539
Encompass_Preferred
4-(Trifluoromethyl)benzoic acid-13C6 is the 13C6-labeled form of 4-(trifluoromethyl)benzoic acid, supplied as a high-purity solid for analytical and research use. It is intended as a stable isotope tracer and internal standard for quantitative methods including NMR, GC-MS, and LC-MS, and is provided in small quantities with specified storage conditions.
  • High isotopic enrichment for accurate tracer studies.
  • High chemical purity (98.3%).
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS quantitation.
  • White to off-white solid form for easy handling and weighing.
  • Available in small research-scale sizes with recommended cold storage.
Catalog No. 50-003-02539 Supplier Medchemexpress LLC Supplier No. HYW015198S1MG
4-(Trifluoromethyl)benzoic acid-13C6 is the 13C6-labeled form of 4-(trifluoromethyl)benzoic acid, supplied as a high-purity solid for analytical and research use. It is intended as a stable isotope tracer and internal standard for quantitative methods including NMR, GC-MS, and LC-MS, and is provided in small quantities with specified storage conditions.
  • High isotopic enrichment for accurate tracer studies.
  • High chemical purity (98.3%).
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS quantitation.
  • White to off-white solid form for easy handling and weighing.
  • Available in small research-scale sizes with recommended cold storage.