Learn More
Medchemexpress LLC 4-(trifluoromethyl)benzoic acid-13C6 | 2731552-41-1 | MFCD00002562 | 98.3% | 196.08 | C213C6H5F3O2 | 5 MG
SDP
Catalog No. 5000303374
Encompass_Preferred
4-(Trifluoromethyl)benzoic acid-13C6 is the 13C6 isotopically labeled analogue of 4-(trifluoromethyl)benzoic acid supplied as a solid stable-isotope standard for analytical and tracer applications. It is designed for use in quantitative mass spectrometry, method development, and metabolic tracing to improve measurement accuracy and reproducibility.
  • Isotopically labeled with six 13C atoms for clear mass differentiation.
  • High purity (98.3%) suitable for analytical applications.
  • White to off-white solid for easy handling and storage.
  • Available in multiple small-scale sizes for method development.
  • Supplied with certificate of analysis and safety data sheet for quality assurance.
Catalog No. 50-003-03374 Supplier Medchemexpress LLC Supplier No. HYW015198S5MG
4-(Trifluoromethyl)benzoic acid-13C6 is the 13C6 isotopically labeled analogue of 4-(trifluoromethyl)benzoic acid supplied as a solid stable-isotope standard for analytical and tracer applications. It is designed for use in quantitative mass spectrometry, method development, and metabolic tracing to improve measurement accuracy and reproducibility.
  • Isotopically labeled with six 13C atoms for clear mass differentiation.
  • High purity (98.3%) suitable for analytical applications.
  • White to off-white solid for easy handling and storage.
  • Available in multiple small-scale sizes for method development.
  • Supplied with certificate of analysis and safety data sheet for quality assurance.