Learn More
Medchemexpress LLC 4-(Trifluoromethyl)nicotinic acid | 158063-66-2 | 99.5% | C7H4F3NO2 | 100 G
SDP
Supplier:  Medchemexpress LLC HY66035100G
Encompass_Preferred
4-(Trifluoromethyl)nicotinic acid is a metabolite of the pyridinecarboxamide insecticide Flonicamid. It is suitable for research use.
  • Solid form
  • Light yellow to light brown color
  • High purity
Catalog No. 50-004-33801