Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-(Trifluoromethyl)nicotinic acid | 158063-66-2 | 99.5% | C7H4F3NO2 | 25 G
SDP

Catalog No. 5000358533
Encompass_Preferred

4-(Trifluoromethyl)nicotinic acid is a chemical compound primarily known as a metabolite of the insecticide Flonicamid. This product is designed for research use only.

  • Solid appearance, light yellow to light brown in color
  • Purity of 99.53%
  • Store at room temperature for up to 3 years
  • When in solvent, store at -80°C for 2 years or -20°C for 1 year

Catalog No. 50-003-58533 Supplier Medchemexpress LLC Supplier No. 25GHY66035
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-(Trifluoromethyl)nicotinic acid is a chemical compound primarily known as a metabolite of the insecticide Flonicamid. This product is designed for research use only.

  • Solid appearance, light yellow to light brown in color
  • Purity of 99.53%
  • Store at room temperature for up to 3 years
  • When in solvent, store at -80°C for 2 years or -20°C for 1 year

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.