Learn More
Medchemexpress LLC 4-(Trifluoromethyl)nicotinic acid | 158063-66-2 | 99.5% | C7H4F3NO2 | 500 G
SDP
Catalog No. 5000433943
Encompass_Preferred
4-(Trifluoromethyl)nicotinic acid is a metabolite of the insecticide Flonicamid. This compound is intended for research use only.
  • Shipped at room temperature
  • Store at room temperature for 3 years
  • In solvent, store at -80°C for 2 years
  • In solvent, store at -20°C for 1 year
Catalog No. 50-004-33943 Supplier Medchemexpress LLC Supplier No. HY66035500G
4-(Trifluoromethyl)nicotinic acid is a metabolite of the insecticide Flonicamid. This compound is intended for research use only.
  • Shipped at room temperature
  • Store at room temperature for 3 years
  • In solvent, store at -80°C for 2 years
  • In solvent, store at -20°C for 1 year