Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[4-(Trifluoromethyl)phenyl](2,4,6-trimethylphenyl)iodonium Trifluoromethanesulfonate 98.0+%, TCI America™
SDP

Catalog No. T34461G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T34461G 1 g
T34465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T34461G Supplier TCI America Supplier No. T34461G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1232133-62-8
Molecular Formula C17H15F6IO3S
Molecular Weight (g/mol) 540.259
MDL Number MFCD27978254
InChI Key XTTMIKRDUJQRGJ-UHFFFAOYSA-M
Synonym Mesityl[4-(trifluoromethyl)phenyl]iodonium Triflate
PubChem CID 117064601
IUPAC Name trifluoromethanesulfonate;[4-(trifluoromethyl)phenyl]-(2,4,6-trimethylphenyl)iodanium
SMILES CC1=CC(=C(C(=C1)C)[I+]C2=CC=C(C=C2)C(F)(F)F)C.C(F)(F)(F)S(=O)(=O)[O-]

Specifications

Melting Point 203°C
Quantity 1 g
Formula Weight 540.26
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material [4-(Trifluoromethyl)phenyl](2,4,6-trimethylphenyl)iodonium Trifluoromethanesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.