Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Trifluoromethyl)phenyl isothiocyanate, 98+%

Catalog No. AAL1189003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1189003 1 g
AAL1189006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1189003 Supplier Thermo Scientific Chemicals Supplier No. L1189003
Add to Cart

CAS: 1645-65-4 | C8H4F3NS | 203.182 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1645-65-4
Molecular Formula C8H4F3NS
Molecular Weight (g/mol) 203.182
MDL Number MFCD00039645
InChI Key DQEVDFQAYLIBRD-UHFFFAOYSA-N
Synonym 4-trifluoromethyl phenyl isothiocyanate, 1-isothiocyanato-4-trifluoromethyl benzene, 4-trifluoromethyl phenylisothiocyanate, 4-trifluoromethylphenylisothiocyanate, p-trifluoromethylphenyl isothiocyanate, benzene, 1-isothiocyanato-4-trifluoromethyl, alpha,alpha,alpha-trifluoro-p-tolyl isothiocyanate, isothiocyanic acid 4-trifluoromethyl phenyl ester, 4-trifluoromethylphenyl isothiocyanate, 1-isothiocyanato-4-trifluoromethyl-benzene
PubChem CID 137134
IUPAC Name 1-isothiocyanato-4-(trifluoromethyl)benzene
SMILES C1=CC(=CC=C1C(F)(F)F)N=C=S

Specifications

Melting Point 39°C to 42°C
Boiling Point 207°C
Flash Point >110°C (230°F)
Odor Pungent
Quantity 1 g
UN Number UN2923
Beilstein 2693553
Sensitivity Moisture sensitive
Formula Weight 203.19
Percent Purity ≥98%
Chemical Name or Material 4-(Trifluoromethyl)phenyl isothiocyanate
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.