Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Trifluoromethyl)phenylacetonitrile 98.0+%, TCI America™
SDP

Catalog No. T23685G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T23685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T23685G Supplier TCI America Supplier No. T23685G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2338-75-2
Molecular Formula C9H6F3N
Molecular Weight (g/mol) 185.15
MDL Number MFCD00001921
InChI Key QNKOCFJZJWOXDE-UHFFFAOYSA-N
Synonym 4-trifluoromethyl phenylacetonitrile, 2-4-trifluoromethyl phenyl acetonitrile, 4-trifluoromethyl benzyl cyanide, p-trifluoromethylbenzyl cyanide, 4-trifluoromethyl phenyl acetonitrile, 4-trifluoromethyl-phenyl-acetonitrile, benzeneacetonitrile, 4-trifluoromethyl, 4-trifluoromethylbenzyl cyanide, p-trifluoromethylphenylacetonitrile
PubChem CID 75359
IUPAC Name 2-[4-(trifluoromethyl)phenyl]acetonitrile
SMILES FC(F)(F)C1=CC=C(CC#N)C=C1

Specifications

Melting Point 47°C
Color White-Yellow
Boiling Point 132°C
Quantity 5 g
UN Number 3439
Formula Weight 185.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Trifluoromethyl)phenylacetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.