Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Trifluoromethyl)salicylic Acid 98.0+%, TCI America™
SDP

Catalog No. T206425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T206425G 25 g
T20645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T206425G Supplier TCI America Supplier No. T206425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 328-90-5
Molecular Formula C8H5F3O3
Molecular Weight (g/mol) 206.12
MDL Number MFCD01529657
InChI Key XMLFPUBZFSJWCN-UHFFFAOYSA-N
Synonym 2-Hydroxy-4-(trifluoromethyl)benzoic Acid
PubChem CID 164578
IUPAC Name 2-hydroxy-4-(trifluoromethyl)benzoic acid
SMILES OC(=O)C1=CC=C(C=C1O)C(F)(F)F

Specifications

Melting Point 178°C
Color White
Quantity 25 g
Formula Weight 206.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Trifluoromethyl)salicylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.