Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Trifluoromethyl)styrene, 98%, stab.

Catalog No. AAH5351303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5351303 1 g
AAH5351306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5351303 Supplier Thermo Scientific Chemicals Supplier No. H5351303
Only null left
Add to Cart

CAS: 402-50-6 | C9H7F3 | 172.15 g/mol

4-(Trifluoromethyl)styrene is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Trifluoromethyl)styrene is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Moisture & light sensitive. Incompatible with oxidizing agents, acids, bases, heat, ultravioloet radiation and free radical initiators.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 402-50-6
Molecular Formula C9H7F3
Molecular Weight (g/mol) 172.15
MDL Number MFCD00075539
InChI Key CEWDRCQPGANDRS-UHFFFAOYSA-N
Synonym 1-trifluoromethyl-4-vinylbenzene, 4-trifluoromethyl styrene, 4-trifluoromethylstyrene, 1-ethenyl-4-trifluoromethyl benzene, 4-vinylbenzotrifluoride, 1-trifluoromethyl-4-vinyl-benzene, 4-trifluoromethystyrene, p-trifluoromethylstyrene, pubchem15485, acmc-1aivr
PubChem CID 2782798
IUPAC Name 1-ethenyl-4-(trifluoromethyl)benzene
SMILES C=CC1=CC=C(C=C1)C(F)(F)F

Specifications

Density 1.165
Boiling Point 65°C to 66°C (40 mmHg)
Flash Point 42°C (108°F)
Refractive Index 1.466
Quantity 1 g
UN Number UN1993
Beilstein 1863548
Sensitivity Light and moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 172.15
Percent Purity 98%
Chemical Name or Material 4-(Trifluoromethyl)styrene, stabilized
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.