Learn More
Medchemexpress LLC 4-(Trifluoromethyl)umbelliferone | 575-03-1 | 98.7% | 10 G
SDP
Catalog No. 5000411044
Encompass_Preferred
4-(Trifluoromethyl)umbelliferone is a fluorescent probe substrate for rat hepatic cytochrome P450 enzymes.
  • Fluorescent probe substrate for rat hepatic cytochrome P450 enzymes
  • Fluorescent dye
  • Enzyme labeling
  • Excitation wavelength (Ex): 335
  • Emission wavelength (Em): 460
  • Protease substrate
  • Ex <380 nm ultraviolet
  • Em 451-495 nm blue
Catalog No. 50-004-11044 Supplier Medchemexpress LLC Supplier No. HYD014910G
4-(Trifluoromethyl)umbelliferone is a fluorescent probe substrate for rat hepatic cytochrome P450 enzymes.
  • Fluorescent probe substrate for rat hepatic cytochrome P450 enzymes
  • Fluorescent dye
  • Enzyme labeling
  • Excitation wavelength (Ex): 335
  • Emission wavelength (Em): 460
  • Protease substrate
  • Ex <380 nm ultraviolet
  • Em 451-495 nm blue