Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-Trifluoromethylsalicylic acid | 328-90-5 | MFCD01529657 | 99.9% | 206.12 | C8H5F3O3 | 1 ML
SDP

Catalog No. 5000302923
Encompass_Preferred

4-Trifluoromethylsalicylic acid (desacetyl triflusal) is an active metabolite of triflusal that functions as a platelet aggregation inhibitor. The compound is supplied as a research-grade analytical standard in solid form and as a ready-to-use solution for biochemical assays.

  • High purity (99.93%) suitable for analytical applications.
  • Molecular formula C8H5F3O3 and molecular weight 206.12.
  • Supplied as solid and as a 10 mM solution in DMSO for convenience.
  • Suitable as a reference standard for assay development and platelet function studies.
  • Registered under CAS 328-90-5 for unambiguous identification.

Catalog No. 50-003-02923 Supplier Medchemexpress LLC Supplier No. HYW01463210MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

4-Trifluoromethylsalicylic acid (desacetyl triflusal) is an active metabolite of triflusal that functions as a platelet aggregation inhibitor. The compound is supplied as a research-grade analytical standard in solid form and as a ready-to-use solution for biochemical assays.

  • High purity (99.93%) suitable for analytical applications.
  • Molecular formula C8H5F3O3 and molecular weight 206.12.
  • Supplied as solid and as a 10 mM solution in DMSO for convenience.
  • Suitable as a reference standard for assay development and platelet function studies.
  • Registered under CAS 328-90-5 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.