Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Triphenylmethylphenol 98.0+%, TCI America™
SDP

Catalog No. T16545G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T16545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T16545G Supplier TCI America Supplier No. T16545G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 978-86-9
Molecular Formula C25H20O
Molecular Weight (g/mol) 336.43
MDL Number MFCD00002364
InChI Key NIPKXTKKYSKEON-UHFFFAOYSA-N
Synonym 4-triphenylmethylphenol, 4-triphenylmethyl phenol, p-tritylphenol, p-triphenylmethylphenol, phenol, 4-triphenylmethyl, p-cresol, .alpha.,.alpha.,.alpha.-triphenyl, 4-tritylphenol, p-triphenylmethyl phenol, 4-tri phenyl methyl phenol, phenol,4-triphenylmethyl
PubChem CID 70422
IUPAC Name 4-(triphenylmethyl)phenol
SMILES OC1=CC=C(C=C1)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 5 g
Formula Weight 336.43
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Triphenylmethylphenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.