Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Undecanone 95.0+%, TCI America™
SDP

Catalog No. U004110ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U004110ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. U004110ML Supplier TCI America Supplier No. U004110ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 14476-37-0
Molecular Formula C11H22O
Molecular Weight (g/mol) 170.30
MDL Number MFCD00048923
InChI Key NBSLHMOSERBUOV-UHFFFAOYSA-N
Synonym 4-undecanone, heptyl propyl ketone, n-heptyl n-propyl ketone, acmc-1c992
PubChem CID 84467
IUPAC Name undecan-4-one
SMILES CCCCCCCC(=O)CCC

Specifications

Color Yellow
Boiling Point 107°C
Quantity 10 mL
Formula Weight 170.30
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Undecanone
EINECSNumber (2)-2475
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.