Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Vinyl-1,3-dioxolan-2-one

Catalog No. 496820100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
5 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-682-0100G 100 g
49-682-01KG 1 kg
49-682-05KG 5 kg
49-682-025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 49-682-0100G Supplier Sigma Aldrich Supplier No. 496820100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 4-vinyl-1,3-dioxolan-2-one (vec) is a cyclic carbonate, that can be synthesized from acrolein by reacting with sulfur ylide and co2
  • The ruthenium-catalyzed transfer hydrogenation (th) of vec in the presence of 2-propanol forms 1,2-butanediol
  • Indolines and indoles undergo selective c-h allylation with vec in the presence of rhodium(iii) catalyst.used to synthesize functional polymers

Specifications

CAS 4427-96-7
Density 1.188 g/mL (at 25°C (literature))
Boiling Point 237°C (733 mmHg)
Molecular Formula C5H6O3
Refractive Index n20/D 1.45 (literature)
Linear Formula C5H6O3
MDL Number MFCD00143315
Quantity 100 g
Synonym VEC; Vinyl ethylene carbonate
Molecular Weight (g/mol) 114.1
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.