Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Vinylaniline

Catalog No. 5361805G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-618-05G 5 g
53-618-01G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 53-618-05G Supplier Sigma Aldrich Supplier No. 5361805G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4-Vinylaniline (4-VAn) is a primary amine surfactant.4-VAn undergoes graft copolymerization with poly(tetrafluoroethylene) (PTFE) and Si surface, followed by oxidative copolymerization with aniline. Thus, it renders PTFE and Si surface conductive. 4-VAn is coupled with hydrogen terminated Si surfaces for electroless metal and synthetic metal deposition. Palladium(II) schiff base complexes derived from Allylamine and vinylaniline has been reported. It also acts as a second surfactant for coating nanomagnetic particles. It is used in functionalization of single-walled carbon nanotube through solvent free functionalization.

Specifications

CAS 1520-21-4
Density 1.017 g/mL (at 25°C)
Boiling Point 213°C to 214°C
Molecular Formula C8H9N
Refractive Index n20/D 1.626 (literature)
Linear Formula H2C=CHC6H4NH2
Quantity 5 g
MDL Number MFCD00015329
Synonym 4-Aminostyrene
Molecular Weight (g/mol) 119.16
Percent Purity 97%
Biological Activity Respiratory system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.