Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Vinylbenzoic acid, 96%

Catalog No. AC294910050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC294910050 5 g Glass bottle
AC294910010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC294910050 Supplier Thermo Scientific Chemicals Supplier No. 294910050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1075-49-6 | C9H8O2 | 148.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1075-49-6
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.16
MDL Number MFCD00002569
InChI Key IRQWEODKXLDORP-UHFFFAOYSA-N
Synonym 4-vinylbenzoic acid, p-vinylbenzoic acid, 4-carboxystyrene, benzoic acid, 4-ethenyl, styrene-4-carboxylic acid, 4-vinyl-benzoic acid, p-vinyl benzoic acid, 4-vinyl benzoic acid, para-vinyl benzoic acid, pubchem12624
PubChem CID 14098
IUPAC Name 4-ethenylbenzoic acid
SMILES C=CC1=CC=C(C=C1)C(=O)O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 142.0°C to 144.0°C
Color Beige to White
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Linear Formula CH2CHC6H4COOH
Quantity 5 g
Solubility Information (10% in Methanol) Clear solution
Stabilizer 0.1% t-butylcatechol
Formula Weight 148.16
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 4-Vinylbenzoic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 214-053-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.