Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Vinylbiphenyl, 97%

Catalog No. AAL1186903
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1186903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1186903 Supplier Thermo Scientific Chemicals Supplier No. L1186903
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2350-89-2 | C14H12 | 180.25 g/mol

4-Vinylbiphenyl intermediate is used in organic synthesis and in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Vinylbiphenyl intermediate is used in organic synthesis and in chemical research.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilat
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2350-89-2
Molecular Formula C14H12
Molecular Weight (g/mol) 180.25
MDL Number MFCD00008620
InChI Key HDBWAWNLGGMZRQ-UHFFFAOYSA-N
Synonym 4-vinylbiphenyl, p-vinylbiphenyl, 4-phenylstyrene, biphenyl, 4-vinyl, 1,1'-biphenyl, 4-ethenyl, p-phenylstyrene, 4-ethenyl-1,1'-biphenyl, 4-vinyldiphenyl, 4-vinyl biphenyl, 4-vinyl-biphenyl
PubChem CID 16883
IUPAC Name 1-ethenyl-4-phenylbenzene
SMILES C=CC1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 115°C to 116°C
Quantity 1 g
Beilstein 2039221
Solubility Information Insoluble in water.
Formula Weight 180.25
Percent Purity 97%
Chemical Name or Material 4-Vinylbiphenyl
EINECSNumber 219-082-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.