Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-vinylcyclohexene dioxide | 106-87-6 | MFCD00022354 | 98.0% | 140.18 g/mol | C8H12O2 | 1 G
SDP

Catalog No. 5000240966
Encompass_Preferred

4-Vinylcyclohexene dioxide is a diepoxide research reagent used to study reproductive toxicity and premature ovarian insufficiency. It is a colorless to light yellow liquid with molecular formula C8H12O2, molecular weight 140.18 g/mol, and reported purity 98.0% (NMR). Handle and store per supplier recommendations; material is for research use only and may be restricted in some territories.

  • Used to model ovarian follicle toxicity and reproductive toxicity
  • Colorless to light yellow liquid suitable for laboratory use
  • Purity 98.0% (NMR) as reported by certificate of analysis
  • Molecular weight 140.18 g/mol; formula C8H12O2
  • Storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months)
  • For research use only; not for human or clinical use

Catalog No. 50-002-40966 Supplier Medchemexpress LLC Supplier No. HY106591A1G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Vinylcyclohexene dioxide is a diepoxide research reagent used to study reproductive toxicity and premature ovarian insufficiency. It is a colorless to light yellow liquid with molecular formula C8H12O2, molecular weight 140.18 g/mol, and reported purity 98.0% (NMR). Handle and store per supplier recommendations; material is for research use only and may be restricted in some territories.

  • Used to model ovarian follicle toxicity and reproductive toxicity
  • Colorless to light yellow liquid suitable for laboratory use
  • Purity 98.0% (NMR) as reported by certificate of analysis
  • Molecular weight 140.18 g/mol; formula C8H12O2
  • Storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months)
  • For research use only; not for human or clinical use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.