Learn More
Selleck Chemical LLC 4-Vinylcyclohexene dioxide
SDP
Supplier:  Selleck Chemical LLC E00241G
Encompass_Preferred
4-Vinylcyclohexene dioxide (Vinylcyclohexene dioxide VCD) is a chemical used to induce menopause and decrease estrogen production 4-Vinylcyclohexene dioxide is used as a crosslinking agent for the production of epoxy resins
Catalog No. 50-313-6064