Learn More
Selleck Chemical LLC 4-Vinylcyclohexene dioxide
SDP
Catalog No. 503136668
Encompass_Preferred
4-Vinylcyclohexene dioxide (Vinylcyclohexene dioxide VCD) is a chemical used to induce menopause and decrease estrogen production 4-Vinylcyclohexene dioxide is used as a crosslinking agent for the production of epoxy resins
Catalog No. 50-313-6668 Supplier Selleck Chemical LLC Supplier No. E0024100MG
4-Vinylcyclohexene dioxide (Vinylcyclohexene dioxide VCD) is a chemical used to induce menopause and decrease estrogen production 4-Vinylcyclohexene dioxide is used as a crosslinking agent for the production of epoxy resins