Learn More
Medchemexpress LLC 4-Vinylphenol | 2628-17-3 | 120.15 | 1 ML
SDP
Catalog No. 5000445546
Encompass_Preferred
4-Vinylphenol is a compound found in the medicinal herb Hedyotis diffusa Willd and wild rice. It is also a metabolite of p-coumaric and ferulic acid produced by lactic acid bacteria in wine. This compound induces apoptosis, inhibits blood vessel formation, and suppresses invasive breast tumor growth in vivo.
  • Significantly reduces cell viability in parental MDA-MB-231 cells, with an IC50 value of 109 μg/mL.
  • Reduces sphere formation and vimentin expression on CSCs in breast cancer.
  • Significantly suppresses cell proliferation in CSC-enriched MDA-MB-231 cells.
  • Significantly reduces ALDH1A1 expression by 50% in CSC-enriched MDA-MB-231 cells.
  • Significantly increases the expressions of caspase 3, likely sensitizing CSC-enriched MDAMB-231 cells to apoptosis.
Catalog No. 50-004-45546 Supplier Medchemexpress LLC Supplier No. HYW00528810MM/1ML
4-Vinylphenol is a compound found in the medicinal herb Hedyotis diffusa Willd and wild rice. It is also a metabolite of p-coumaric and ferulic acid produced by lactic acid bacteria in wine. This compound induces apoptosis, inhibits blood vessel formation, and suppresses invasive breast tumor growth in vivo.
  • Significantly reduces cell viability in parental MDA-MB-231 cells, with an IC50 value of 109 μg/mL.
  • Reduces sphere formation and vimentin expression on CSCs in breast cancer.
  • Significantly suppresses cell proliferation in CSC-enriched MDA-MB-231 cells.
  • Significantly reduces ALDH1A1 expression by 50% in CSC-enriched MDA-MB-231 cells.
  • Significantly increases the expressions of caspase 3, likely sensitizing CSC-enriched MDAMB-231 cells to apoptosis.