Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Vinylpyridine

Catalog No. 503125717 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-312-5717 100 mL
50-312-5718 500 mL
50-312-5871 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-312-5717 Supplier Sigma Aldrich Supplier No. V3204100ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Polymerization monomer and protein alkylating agent.

Specifications

CAS 100-43-6
Density 0.975 g/mL (at 25°C)
Boiling Point 62°C to 65°C (15 mmHg)
Molecular Formula C7H7N
Refractive Index n20/D 1.549 (literature)
Linear Formula C7H7N
MDL Number MFCD00006447
Quantity 100 mL
Molecular Weight (g/mol) 105.14
Percent Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.