Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Vinylpyridine, 96%, low polymer, stab. with 100ppm hydroquinone

Catalog No. AAH36621AE
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH36621AE 100 mL
AAH36621AC 25 mL
AAH36621AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH36621AE Supplier Thermo Scientific Chemicals Supplier No. H36621AE
Only null left
Add to Cart

CAS: 100-43-6 | C7H7N | 105.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-43-6
Molecular Formula C7H7N
Molecular Weight (g/mol) 105.14
MDL Number MFCD00006447
InChI Key KFDVPJUYSDEJTH-UHFFFAOYSA-N
Synonym 4-vinylpyridine, pyridine, 4-ethenyl, pyridine, 4-vinyl, gamma-vinylpyridine, unii-i56g67xm8d, ccris 5240, 4vp, 4-pyridylethylene, 4-vinyl-pyridine, 4-vinyl pyridine
PubChem CID 7502
IUPAC Name 4-ethenylpyridine
SMILES C=CC1=CC=NC=C1

Specifications

Density 0.984
Boiling Point 61°C (12 mmHg)
Flash Point 51°C (124°F)
Refractive Index 1.5512
Quantity 100 mL
UN Number UN3073
Formula Weight 105.14
Percent Purity 96%
Chemical Name or Material 4-Vinylpyridine, stabilized with 100ppm hydroquinone
Hazard Category H226-H301+H311+H331-H314-H317-H335
Hazard Statement GHS H Statement
H301-H331-H334-H314-H318-H226-H317
Toxic if swallowed.
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
May cause an allergic skin reaction.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P272-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: VINYLPYRIDINES, STABILIZED
EINECSNumber 202-852-0
RTECSNumber UU1045000
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.