Learn More
Kuhner Shaker Inc 4 WATER TRAPS RECOMMEND FOR US
Catalog No. 502570078
Encompass
4 water traps recommend for use with Baffled Flasks..Use with TOM GL45 screw caps
Catalog No. 50-257-0078 Supplier Kuhner Shaker Inc Supplier No. SMTO00030
4 water traps recommend for use with Baffled Flasks..Use with TOM GL45 screw caps