Learn More
Genesee Scientific Corporation 4-Way Flipper Microtube Rack with Tray, 4/pk
SDP
Catalog No. NC1874274
Encompass
This assorted four-sided rack provides a great long-lasting alternative to cardboard freezer boxes. Made of durable polypropylene, the 4-Way Flipper Rack is guaranteed to keep its shape and withstand freezing and autoclaving conditions. Each side of the rack is designed to accommodate tube sizes from 0.5ml to 50ml. For easy identification, the outskirts of the rack are labeled with easy-to-read letters and numbers.
Catalog No. NC1874274 Supplier Genesee Scientific Corporation Supplier No. 27217
This assorted four-sided rack provides a great long-lasting alternative to cardboard freezer boxes. Made of durable polypropylene, the 4-Way Flipper Rack is guaranteed to keep its shape and withstand freezing and autoclaving conditions. Each side of the rack is designed to accommodate tube sizes from 0.5ml to 50ml. For easy identification, the outskirts of the rack are labeled with easy-to-read letters and numbers.