Learn More
Research Products International Corp 4-Way Joiner for Modular Beta Workstation
SDP
Catalog No. 501978086
Encompass
Create the ideal size beta workstation with our unique modular system. Simply slide 3/8 inch thick acrylic panels into upright joiners to form any size enclosure or barrier without using tools. System can be easily disassembled for convenient storage when not in use.
All components available separately in the standard sizes listed. Custom sizes can also be supplied, please inquire.
Catalog No. 50-197-8086 Supplier Research Products International Corp Supplier No. BR216
Create the ideal size beta workstation with our unique modular system. Simply slide 3/8 inch thick acrylic panels into upright joiners to form any size enclosure or barrier without using tools. System can be easily disassembled for convenient storage when not in use.
All components available separately in the standard sizes listed. Custom sizes can also be supplied, please inquire.